Learn More
Grainger Conductive Elec Tape 18 Lx1/4 W CFL-5A
Supplier:  Grainger 15D557
Encompass_Preferred
The TAPECASE Conductive Elec Tape 18" L x 1/4" W CFL-5A is an 18 inch long by 1/4 inch wide conductive aluminum foil tape with conductive acrylic adhesive. It has a temperature range of -40°F to 300°F and is UL listed. This tape provides EMI/RFI shielding and can be used for aircraft, medical and electronic applications. It has a peel strength of 35 oz/in and tensile strength of 22 lb/in. The tape comes in a convenient dispenser box for easy use.
Catalog No. 50-274-8749