Learn More
Grainger Conductive Elec Tape 18 Lx1 W CFL-5A
Supplier:  Grainger 15D560
Encompass_Preferred
The TAPECASE Conductive Electrical Tape 18" Length x 1" Width CFL-5A is a conductive aluminum foil tape with conductive acrylic adhesive. It has a thickness of 3.5 mils and is UL listed. This tape is designed for EMI/RFI shielding, grounding, and static charge/discharge applications. It can be applied to metals, plastics, glass, and more. The operating temperature range is -40°F to 300°F. This 18" x 1" roll of tape comes in a dispenser box for easy use.
Catalog No. 50-275-2145