Learn More
Grainger Conductive Elec Tape 18 Lx2 W CFL-5A
Supplier:  Grainger 15D561
Encompass_Preferred
The TAPECASE Conductive Electrical Tape is a self-adhesive, conductive tape used for EMI/RFI shielding, grounding, and static charge dissipation. It has a conductive acrylic adhesive and is UL-listed. This black tape comes in multiple widths (1/2", 3/4", 1") on a standard 60 yard roll. It has a temperature rating of up to 221°F and is resistant to moisture, chemicals, and UV rays. The tape has a surface resistivity of less than 0.05 ohms and conforms to irregular surfaces. It can be applied to metals, plastics, glass, and more. This is an industrial-grade electrical tape ideal for a variety of EMI/RFI shielding and grounding applications.
Catalog No. 50-275-2146