Learn More
Grainger Conductive Elec Tape 18 Lx3/4 W CFL-5A
Supplier:  Grainger 15D559
Encompass_Preferred
The TAPECASE Conductive Electrical Tape is a self-adhesive, conductive tape used for EMI/RFI shielding, grounding, and static charge dissipation. It has a conductive acrylic adhesive and is UL-listed. This black tape comes in multiple widths (1/2", 3/4", 1") on a standard 60 yard roll. It has a temperature range of -30°F to 200°F and insulation resistance of 0.05 ohms. The tape has high shielding effectiveness across a wide frequency range. It conforms well to irregular surfaces and has good aging properties. This is a quality electrical tape for EMI shielding, grounding, and static dissipation applications.
Catalog No. 50-275-1153