Learn More
Grainger Conductive Elec Tape 18 Lx3 W CFL-5A
Supplier:  Grainger 15D562
Encompass_Preferred
The TapeCase Conductive Electrical Tape is a self-adhesive, conductive tape used for EMI/RFI shielding, grounding, and static charge dissipation. It has a conductive acrylic adhesive and is UL-listed. This black tape comes in several widths (1/4", 1/2", 1") on a standard 60 yard roll. It has a temperature range of -20°F to 200°F and insulation resistance of 0.05 ohms. The tape has high shielding effectiveness across a wide frequency range. It conforms well to irregular surfaces and forms a strong bond for permanent applications. This tape can shield cables, provide grounding, and prevent static buildup in electronics and electrical applications.
Catalog No. 50-275-2595