Learn More
Hach Company Contacting Conductivity Sensor for Low Conductivity, K=0.5, 3/4" Kynar Compression Fitting
Supplier:  Hach Company 3422B3A
Encompass
Nominal Cell Constant (K) = 0.5, 3/4 inch NPT Kynar (PVDF) Compression Fitting, 6m (20ft) Analog Cable, Titanium Electrode
Catalog No. 50-199-7438