Learn More
Medchemexpress LLC HY-100944 5mg Medchemexpress, Conduritol B epoxide CAS:6090-95-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1009445MG
Encompass_Preferred
Medchemexpress, HY-100944 5mg Conduritol B epoxide CAS:6090-95-5 Conduritol B epoxide is an irreversible covalently bound acid β-glucosidase (GCase) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7791