Learn More
Cayman Chemical ConglobatIn C1 5mg
SDP
Supplier:  Cayman Chemical 332565 MG
Encompass_Preferred
A bacterial metabolite; cytotoxic to NS-1 mouse myeloma cells (IC50 = 1.05 UG/ml)
Catalog No. 50-294-7952