Learn More
Selleck Chemical LLC Congo Red
SDP
Supplier:  Selleck Chemical LLC S606525MG
Encompass_Preferred
Congo Red (Direct Red 28) is an indicator dye that is blue-violet at pH 3 0 and red at pH 5 0 It is an acid dye used in testing for hydrochloric acid in gastric contents but has carcinogenic properties
Catalog No. 50-000-04286