Learn More
Polycarbin Reversible Conical and Microcentrifuge Tube Rack, 4 Way Flipper
SDP
Supplier:  Polycarbin 2PR04
Encompass_Preferred
Polycarbin's 4-Way Flipper Tube Rack is designed for sustainability-minded scientists seeking a versatile solution for benchtop work or cold storage. Made from 100% recycled content, this durable polypropylene rack features a compact, four-sided design that accommodates a variety of tube sizes, including 32 x 0.5 mL tubes, 32 x 1.5–2.0 mL tubes, 12 x 5–15 mL tubes, and 4 x 50 mL tubes. Autoclavable and resistant to common lab chemicals, it provides a reliable and eco-friendly alternative for organizing and storing laboratory samples.
Catalog No. NC3683374