Learn More
Chemglass Life Sciences 5mL Conical Reaction Vial, Minum-Ware, Graduated to 4mL
SDP
Supplier:  Chemglass Life Sciences MW8005
Encompass_Preferred
5mL Conical Reaction Vial, Minum-Ware, Graduated to 4mL. Heavy wall vial has an internal conical bottom for optimum sample retrieval and flat base which provides added mechanical strength and bench stability. Vials are graduated at the intervals listed. With 14/10 threaded outer joint. Supplied complete with open-top screw cap (MW-102-33), PTFE faced septa (MW-104-33) and Viton O-ring (MW-100-08).
Catalog No. MW8005