Learn More
Glass Expansion Conikal DC Nebulizer 1mL/min with 1,500mm tubing, Includes NFTS-075 UniFit Connector with 0.75mm ID x 1.3mm OD x 1500mm long tubing (1 piece), and 70-803-1311 Gas Fitting with Ratchet Nebulizer connector
SDP
Supplier:  Glass Expansion A411UC1S
Encompass_Preferred
Conikal DC Nebulizer 1.0mL/min, includes UniFit Connector with 0.75mm ID x 1.3mm OD x 1500mm long tubing (1 pc) and DC Gas Fitting Type 41 with ratchet connector (1 pc). Compatible with Shimadzu 9000/9800 ICP-OES.Optimum nebulizer gas flow rate is 1.0L/min and sample flow rate is 1.0mL/min. Recommended not to exceed optimum 1.0mL/min sample flow rate; however, lower gas and sample flow rates are OK. The backpressure should read approximately 40 psi at 1.0L/min gas flow rate. The Conikal nebulizer is a high-performance, general use glass nebulizer, that tolerates up to 5% TDS and 75um particulates.Replacement parts include P/Ns NFTS-075 and 501907301.Recommend nebulizer maintenance tools are Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7485