Learn More
Raybiotech Inc Conjugation Kit for Super Mag Carboxyl Magnetic Beads (50 nm)
Supplier:  Raybiotech Inc 8CSK50
Encompass_Preferred
Kit Components: Carboxyl Super Mag Magnetic Beads (8CSK-50): 1 mL (10 mg/mL) ; Activation Buffer: 30 mL; Quenching Buffer: 2 mL; Storage Buffer: 50 mL; EDC: 25 mg; Sulfo-NHS: 25 mg.
RayBiotech's carboxyl terminated Super Mag magnetic beads (50 nm, 100 nm, 150 nm and 200 nm) are now available in an easy-to-use kit format to enable researchers to conjugate proteins/ligands of their own choice to these magnetic beads. Attribute to the large surface area, excellent colloidal stability and unique surface coating of the Carboxyl Super Mag (50 nm), the kit exhibits high binding capacity and low non-specific binding. The recommended protocol has been used to successfully conjugate bovine serum albumin, streptavidin or immunoglobulin to these magnetic beads. Shelf Life: 12 months
Catalog No. NC2103992