Learn More
Glass Expansion Connector for Noordermeer Nebulizer (PKT 10)
SDP
Supplier:  Glass Expansion EZT1
Encompass_Preferred
Connector for Noordermeer V-Groove Nebulizer (PKT 10). Packet of 10 replacement sample lines for the Noordermeer V-Groove Nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC2001233