Learn More
AVANTOR MASTERFLEX (VWR LLC) Barbed fittings, Straight Connectors, 1/8"", Polypropylene, 25 pack
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0636522
Encompass
Barbed fittings, Straight Connectors, Polypropylene, 1/8" ID, 25/Pk, Quick and easy assembly without toolsIdeal for low-pressure applicationsChemical resistantAvailable in a variety of sizes, materials, and configurations
Catalog No. NC0894988