Learn More
Med Vet International Dynarex Sterile Oil Emulsion Dressing, 5" X 9", 72/Case
Supplier: Med Vet International 3070
A non-adherent knitted gauze impregnated with a petrolatum and sunflower oil in an emulsion blend. These conformable, non-occlusive dressings allow exudate to flow away from the wound into a secondary dressing, minimizing skin maceration. Knitted gauze dressings, evenly impregnated with an emulsion blend of petrolatum and sunflower oil. Allow exudate to flow away from the wound, minimizing the risk of skin maceration. Non-occlusive, primary dressing used for low exuding wounds at stages 2, 3 or 4.