Learn More
Apexbio Technology LLC Conoidin A 18080-67-6 250mg
SDP
Supplier:  Apexbio Technology LLC C4293250
Encompass_Preferred
Conoidin A (CAS 18080-67-6) is a small-molecule inhibitor targeting peroxiredoxin II (PrxII) It is designed to inhibit PrxII thereby modulating redox signaling pathways and peroxidase activity related to oxidative stress Conoidin A exerts its biological activity primarily through covalent modification of the active-site cysteine residues in PrxII resulting in irreversible inhibition of enzyme function In cellular studies Conoidin A demonstrates inhibitory activity with an IC50 value of approximately 23 M Based on these pharmacological properties Conoidin A holds research potential in the investigation of oxidative stress mechanisms peroxiredoxin-mediated redox signaling and the functional roles of Prx enzymes in mammalian and parasitic models
Catalog No. 50-000-91486