Learn More
Apexbio Technology LLC ω-Conotoxin GVIA 1mg
SDP
Supplier:  Apexbio Technology LLC B50481
Encompass_Preferred
-Conotoxin GVIA (CAS 106375-28-4) is a peptide neurotoxin derived from marine cone snails It acts as a highly selective blocker of N-type voltage-gated calcium channels (Cav2 2) on neuronal membranes inhibiting calcium influx and consequently reducing presynaptic neurotransmitter release Owing to its specificity for Cav2 2 channels -Conotoxin GVIA is frequently employed in neuroscience research to investigate mechanisms of synaptic transmission pain pathway signaling and neural circuit modulation Its use facilitates the analysis of presynaptic function and the development of models for studying pain and neuropharmacological processes
Catalog No. 50-000-88632