Learn More
LAMDA BIOTECH INC Conquest PCR Master Mix - 2
Supplier:  LAMDA BIOTECH INC D911MIX2
Encompass_Preferred
A pre-formulated 2X PCR master mix optimized for challenging templates, including GC-rich sequences. Features direct gel loading capability and enhanced buffer composition for complex PCR applications.
Catalog No. NC3726373