Learn More
LAMDA BIOTECH INC Conquest™ PCR Master Mix - 4
Supplier:  LAMDA BIOTECH INC D911MIX4
Encompass_Preferred
Thermus aquaticus DNA Polymerase (Taq DNA Polymerase) is a thermostable enzyme which can replicate DNA at 74oC and remain active even after incubation at 95oC. It has very strong 5’-3’ DNA polymerase activity with a fast extension rate. Although it lacks any 3’-5’ exonuclease activity (and therefore proofreading ability), Taq DNA polymerase remains one of the most widely used enzymes in molecular biology. Lamda Biotech’s bacterially derived Taq DNA Polymerase is unique for strong activity and low background in DNA amplification. This enzyme is purified from bacteria. Users should be cautious when dealing with bacteria DNA sequences.
Catalog No. 50-204-5763