Learn More
McMaster-Carr McMaster-Carr, Constant Force Springs
Supplier:  McMaster-Carr 9293K12
Encompass
McMaster-Carr offers a wide selection of constant force springs made from materials like stainless steel, music wire, and carbon steel. These springs provide a constant force throughout their deflection range. They are available in various sizes, forces, lengths, and end types like loops or hooks. Constant force springs are commonly used in applications like retractable cord reels, counterbalancing, and tensioning. McMaster-Carr's springs can be ordered individually or in packs. Customization options are available.
Catalog No. NC2832435