Learn More
Grainger Construction Adhesive 3.7 fl oz Tube
Supplier:  Grainger 16NG99
Encompass_Preferred
The ECLECTIC E6000 Construction Adhesive is a versatile, permanent bonding adhesive ideal for both indoor and outdoor applications. It bonds wood, metal, glass, ceramic, tile, concrete, foam and more. This adhesive is waterproof, flexible and dries clear. It comes in a 2.7 oz tube, has a working time of 45 minutes and cures in 24 hours at room temperature. The E6000 adhesive forms strong, long-lasting bonds and can fill gaps up to 1/4 in. It is VOC compliant and made in the USA.
Catalog No. 50-276-4059