Learn More
Quanterix Inc CONSUMABLES KIT FOR SR-X
SDP
Supplier:  Quanterix Inc 103077
Encompass
  • Consumables kit for ultra-sensitive biomarker detection
  • Compatible with Quanterix SR-X system
  • Utilizes Simoa (Single Molecule Array) technology
  • Includes 16 Simoa discs
  • Contains 4 assay plates, each with 96 tests
  • Comes with 4 lids
  • Includes 4 boxes of disposable pipette tips
  • Supports high-precision analysis of protein and nucleic acid biomarkers
  • Biomarker detection sensitivity is ultra-sensitive (1000X higher than standard immunoassays)
  • Workflow is simple 2-step or 3-step
  • Wash buffers are not included
Catalog No. NC1436157