Learn More
Electron Microscopy Sciences Contact Plate, 60x15 mm, 20 ml
SDP
Supplier:  Electron Microscopy Sciences 7069210
Encompass_Preferred
These polystyrene dishes are free from optical distortion. The dish has 16 of 10 x 10 mm square grids, which is marked numerically in one direction and alphabetically in the other. Dishes are packaged 20 per sleeve and are radiation sterile.
Catalog No. 50-190-0274