Learn More
Medchemexpress LLC Contactin-2/CNTN2 M 10ug
SDP
Supplier:  Medchemexpress LLC HYP7422310UG
Encompass_Preferred
Contactin-2/CNTN2 Protein collaborates with transmembrane protein CNTNAP2 crucially organizing axonal domains at nodes of Ranvier This partnership maintains voltage-gated potassium channels at the juxtaparanodal region emphasizing Contactin-2 s pivotal role in the intricate molecular architecture essential for efficient nerve impulse propagation along myelinated axons Contactin-2/CNTN2 Protein Mouse (HEK293 His) is the recombinant mouse-derived Contactin-2/CNTN2 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78687