Learn More
Essendant CONTAINERDELI48OZ200/C
Supplier:  Essendant ECOEPRC48
Encompass_Preferred
These plant-based alternatives make your food look great Send back the waste that comes with old school containers and reach for something different Renewable and compostable PLA is just as clear as your mom s plastic so you ve got a great option for your Grab n Go needs Remember though PLA only works for cold foods so think recycled content if the goods are hot Made with PLA a plant-based renewable resource type of plastic and meet ASTM standards for composability Being clear they re great for displaying cold your favorite cold foods Engineered for strength and stackability Lids included Food Container Type- Base/Lid Combo Material(s)- Corn-Based Plastic Material Type- Plastic Interior Volume Capacity- 48 oz
Catalog No. 50-000-35361