Learn More
Essendant CONTAINERDELI5OZCLR
Supplier:  Essendant FABPK5SC
Encompass_Preferred
Package salads soups and more in flavor- and odor-transfer-proof containers Printable surface provides branding and labeling opportunities Smooth-rolled doubled-over rims enhance rigidity Product line s standardized diameter delivers one-lid-fits-all convenience Microwave-safe polyethylene withstands temperatures of up to 240 F Food Container Type- Base Material(s)- Polypropylene Material Type- Plastic Interior Volume Capacity- 5 oz
Catalog No. 50-000-35181