Learn More
Medchemexpress LLC Contezolid | 1112968-42-9 | 99.37% | 408.33 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY199151MG
Encompass_Preferred
Contezolid is a new, orally active oxazolidinone antibiotic currently being studied for complicated skin and soft tissue infections (cSSTI) caused by resistant Gram-positive bacteria. It significantly reduces the potential for myelosuppression and monoamine oxidase inhibition (MAOI).
  • New and orally active oxazolidinone antibiotic.
  • In study for complicated skin and soft tissue infections (cSSTI) caused by resistant Gram-positive bacteria.
  • Markedly reduces potential for myelosuppression.
  • Markedly reduces potential for monoamine oxidase inhibition (MAOI).
  • Highly potent against all Gram-positive clinical isolates of staphylococci, streptococci, and enterococci, including MDR organisms such as MRSA, methicilline-resistant *Staphylococcus epidermidis* (MRSE), penicillin-resistant Streptococci (PRSP), and VRE.
  • Rapid oral absorption in mouse, rat, and dog.
  • Exhibits no obvious toxicity.
Catalog No. 50-004-03338