Learn More
Medchemexpress LLC Contezolid | 1112968-42-9 | 99.37% | 408.33 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY19915100MG
Encompass_Preferred
Contezolid (MRX-I) is a new, orally active oxazolidinone, currently under study as an antibiotic for complicated skin and soft tissue infections (cSSTI) caused by resistant Gram-positive bacteria. It significantly reduces the potential for myelosuppression and monoamine oxidase inhibition (MAOI).
  • Orally active oxazolidinone
  • Antibiotic for complicated skin and soft tissue infections (cSSTI) caused by resistant Gram-positive bacteria
  • Markedly reduces potential for myelosuppression and monoamine oxidase inhibition (MAOI)
  • Highly potent against all gram-positive clinical isolates, including MDR organisms
  • Rapid oral absorption in mouse, rat, and dog
  • Exhibits no obvious toxicity
Catalog No. 50-004-01806