Learn More
Medchemexpress LLC Continentalic acid | 19889-23-7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN69085MG
Encompass_Preferred
Continentalic acid, derived from Aralia continentalis, demonstrates activity against Staphylococcus aureus. It exhibits minimum inhibitory concentrations (MICs) of approximately 8-16 μg/mL against both Methicillin-susceptible Staphylococcus aureus (MSSA) and Methicillin-resistant Staphylococcus aureus (MRSA) standard strains.
  • Derived from Aralia continentalis.
  • Exhibits antibacterial activity against S. aureus.
  • Effective against methicillin-resistant and methicillin-susceptible S. aureus strains.
  • Purity of 99.72%.
  • Appears as a white to off-white solid.
  • High solubility in DMSO (100 mg/mL).
Catalog No. 50-004-18737