Learn More
Cayman Chemical ContIntalIc AcId 10mg
SDP
Supplier:  Cayman Chemical 3524010 MG
Encompass_Preferred
A diterpenic acid with diverse biological activities; active against MSSA and clinical isolates of MRSA (MICs = 8 and 8-16 UG/ml, respectively); induces apoptosis in U-2932, OCI-LY1, and Ramos (RA 1) B cell lymphoma cells at 200 µM; improves renal function and decreases renal MDA and peroxidase levels, NO production, and neutrophil infiltration in a mouse model of E. coli- and LPS-induced renal injury
Catalog No. 50-291-3515