Learn More
Med Vet International Controlled Iodine Spray, 16Oz
Supplier:  Med Vet International PHIODINESPRAY
Encompass_Preferred
Controlled Iodine Spray is indicated for use in cattle, horses, swine and sheep as a topical antiseptic for use prior to surgical procedures such as castrating and docking; for ringworm and foot rot; for application to the navel of newborn animals; and for minor cuts, teat sores, bruises and abrasions. Compared to Betadine.Products are not always manufacturer specific. Actual product appearance may differ from website image.
Catalog No. 50-218-8424