Learn More
Andwin Scientific All-purpose CONVERTORS POLY LINED towels
SDP
Supplier:  Andwin Scientific 3520
Encompass_Preferred
All-purpose towels are ideal for general laboratory cleanup or protection of cleanroom surfaces and equipment. Soft nonwoven tissue hand towels are reinforced with scrim laminate. Absorbent Drape Towel, Polylined. 17” x 22” White. Case of 100
Catalog No. NC9532037