Learn More
Cayman Chemical Convza blInII BlInIn 10mg
SDP
Supplier:  Cayman Chemical 3094510 MG
Encompass_Preferred
A diterpene with antiulcerogenic activity; decreases gastric mucosal MDA levels and ulcer area in a rat model of pyloric ligation-induced ulcers
Catalog No. 50-299-1798