Learn More
Med Vet International Cool Renewal™, Cryosurgical Kit, 1 Cannister, J1401C, Each
Supplier: Med Vet International JOR1401C
Cryosurgical by Cool Renewal. - Convenient alternative to liquid nitrogen - Spray canister freezes at -70 degrees C (-94 degrees F) - 2 minute procedure - Minimal patient discomfort - Self anesthetizing - Portable - Minimum 5 year shelf life - Cost about $2 per procedure Cool Renewal. Complete Cryosurgical Kit with Accessories includes: - 1 Canister of Cryogen- 65 Freezes - (2) Extender Tubes - Assorted Pack of 60 Disposable, Double Ended Foam Tipped Applicators (20 each, Small, Medium, Large/XL) - Assorted Pack of 25Disposable Plastic Isolation Funnels (10 each, 5mm, 8mm, 12mm, 14mm, 16mm) - (10) Disposable Skin Tag Tweezers (Unique to Cool Renewal!) - Instructions for Use - Patient Instructions Tear Pad - Applicator Practice Pad