Learn More
Electron Microscopy Sciences Cooper Shot 50g 50 GR
SDP
Supplier:  Electron Microscopy Sciences 2669302
Encompass_Preferred
Used in Bodian's Protargol Method for neurological tissue stains: Nerve fiber and endings. See Luna, L.G.,(ed), Manual of Histologic Staining Methods of the AFIP 3rd Edition, McGraw-Hill, NY, c 1968, p.195. or Clark, G., (ed.). Staining Procedures, 3rd Edition, Williams & Wilkens Co., Baltimore, c. 1973, 98.
Catalog No. 50-301-07