Learn More
Medchemexpress LLC Copanlisib | 1032568-63-0 | 480.52 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHY15346
Encompass_Preferred
Copanlisib is a potent, selective, and ATP-competitive pan-class I PI3K inhibitor. It demonstrates over 2,000-fold selectivity against other lipid and protein kinases, with the exception of mTOR, and exhibits superior antitumor activity, making it suitable for cancer research.
  • Potent, selective, and ATP-competitive pan-class I PI3K inhibitor
  • Has IC₅₀s of 0.5 nM for PI3Kα, 0.7 nM for PI3Kδ, 3.7 nM for PI3Kβ, and 6.4 nM for PI3Kγ
  • Exhibits over 2,000-fold selectivity against other lipid and protein kinases (excluding mTOR)
  • Demonstrates superior antitumor activity
  • Induces apoptosis in a subset of tumor cell lines
  • Completely inhibits PI3K-mediated AKT phosphorylation
  • Potently inhibits cell proliferation in various human tumor cell lines
Catalog No. 50-003-64849