Learn More
Selleck Chemical LLC Copanlisib (BAY 80-6946) 5mg 1032568-63-0
SDP
Supplier:  Selleck Chemical LLC S28025MG
Encompass_Preferred
Copanlisib (BAY 80-6946) is a potent pan-class I PI3K with IC50 of 0.5, 3.7, 6.4, and 0.7 nM in cell-free assays for PI3K?/?/?/? , respectively. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2982