Learn More
Medchemexpress LLC Copanlisib dihydrochloride | 1402152-13-9 | C23H30Cl2N8O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15346A10MM/1ML
Encompass_Preferred
Copanlisib dihydrochloride is a potent, selective, and ATP-competitive pan-class I PI3K inhibitor. It demonstrates over 2,000-fold selectivity against other lipid and protein kinases, excluding mTOR, and exhibits superior antitumor activity.
  • Induces apoptosis in BT20 breast cancer cells
  • Inhibits PI3K-mediated AKT phosphorylation in ELT3 cells
  • Potently inhibits cell proliferation in human tumor cell lines
  • Shows robust antitumor activity in athymic nude rats injected with KPL4 tumor cells
Catalog No. 50-003-73190