Learn More
Sigma Aldrich Fine Chemicals Biosciences Copper Assay Kit sufficien
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MAK1271KT
Encompass
Copper is an essential trace element. Copper-containing enzymes play important roles in iron and catecholamine metabolism free radical scavenging and in the synthesis of hemoglobin elastin and collagen. Copper is mainly present in ceruloplasmin in the liver. Low levels of copper have been associated with mental retardation depigmentation anemia hypotonia and scorbutic changes in bone. Levels of copper are key diagnostic indicator of diseases such as Wilsons disease microcytic hypochromic anemia and bone disease due to reduced collagen synthesis.
Catalog No. 50-290-4109