Learn More
Glass Expansion Copper Foil for Ceramic Outer Tube 31-808-3365 (Pack of 12)
SDP
Supplier:  Glass Expansion 318083377
Encompass_Preferred
Copper Foil for Ceramic Outer Tube (PKT 12). Package of 12 copper foils. Compatible with Glass Expansion’s D-Torch ceramic outer tube (501907212, 31-808-3365) for PerkinElmer Optima 2000/4000/5000/7000 DV ICP-OES.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3133