Learn More
Strem, An Ascensus Company CAS# 7787-70-4. 25g. Copper(I) bromide (99.99%-Cu) PURATREM
SDP
Supplier:  Strem, An Ascensus Company 29290725G
Encompass
CAS# 7787-70-4. 25g. Copper(I) bromide (99.99%-Cu) PURATREM. Molecular Weight: 143.45. Molecular Formula: CuBr. Color/form: off-white to pale-green pwdr. Strem# 29-2907. http://www.strem.com/catalog/v/29-2907/
Catalog No. NC1874108