Learn More
Strem, An Ascensus Company CAS# 7681-65-4. 50g. Copper(I) iodide (99.999%-Cu) PURATREM. MFCD00010978
SDP
Supplier:  Strem, An Ascensus Company 29048550G
Encompass
CAS# 7681-65-4. 50g. Copper(I) iodide (99.999%-Cu) PURATREM. MFCD00010978. Molecular Weight: 190.45. Molecular Formula: CuI. Color/form: off-white to pale brown pwdr. Strem# 29-0485. http://www.strem.com/catalog/v/29-0485/
Catalog No. 50-901-11297
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.