Learn More
Medchemexpress LLC Copper(II) oxalate | 814-91-5 | MFCD00150451 | 98.0% | 151.56 g/mol | CuC2O4 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW14073150G
Encompass_Preferred
Copper(II) oxalate (CuC2O4) is an inorganic copper salt used as a reagent in biochemical, materials, and coordination chemistry research. It is supplied as a dry solid with CAS 814-91-5, molecular weight 151.56 g/mol, and a typical research-grade purity of about 98.0%.
  • Suitable for biochemical assays and coordination chemistry studies.
  • Available as a dry solid for easy weighing and storage.
  • High purity (~98.0%) for research applications.
  • Well-characterized: CAS 814-91-5, formula CuC2O4, MW 151.56 g/mol.
  • Commonly used in materials synthesis and catalytic studies.
Catalog No. 50-002-82699