Learn More
Strem, An Ascensus Company CAS# 1317-38-0. 10g. Copper(II) oxide (99.995%-Cu) PURATREM. MFCD00010979
SDP
Supplier:  Strem, An Ascensus Company 29059010G
Encompass
CAS# 1317-38-0. 10g. Copper(II) oxide (99.995%-Cu) PURATREM. MFCD00010979. Molecular Weight: 79.55. Molecular Formula: CuO. Color/form: 80-100 mesh black pwdr. Strem# 29-0590. http://www.strem.com/catalog/v/29-0590/
Catalog No. 50-901-11298