Learn More
Strem, An Ascensus Company CAS# 1317-38-0. 100g. Copper(II) oxide, min. 97% (99+%-Cu). MFCD00010979
SDP
Supplier:  Strem, An Ascensus Company 932943100G
Encompass
CAS# 1317-38-0. 100g. Copper(II) oxide, min. 97% (99+%-Cu). MFCD00010979. Molecular Weight: 79.54. Molecular Formula: CuO. Color/form: black pwdr. Strem# 93-2943. http://www.strem.com/catalog/v/93-2943/
Catalog No. 50-901-14899