Learn More
Enzo Life Sciences Copper(II) Protoporphyrin IX, (25mg)
SDP
Supplier:  Enzo Life Sciences ALX430048M025
Encompass_Preferred
Negative control for the heme oxygenase inhibitor zinc protoporphyrin IX (Prod. No. ALX-430-049). Formula: C34H32CuN4O4. MW: 624.2. Purity: ≥95%. Appearance: Red to brown solid. Solubility: Soluble in pyridine, dimethyl formamide or DMSO. Colloidal in aqueous base;  slightly soluble in aqueous base/organic solvent mixtures. Use/Stability: Stable for at least 3 years after receipt when stored at -20°C. Product may be unstable in solution if exposed to light. Long Term Storage: -20°C
Catalog No. NC1355611