Learn More
Strem, An Ascensus Company CAS# 7758-98-7. 250g. Copper(II) sulfate, anhydrous, 98%. MFCD00010981
SDP
Supplier:  Strem, An Ascensus Company 932957250G
Encompass
CAS# 7758-98-7. 250g. Copper(II) sulfate, anhydrous, 98%. MFCD00010981. Molecular Weight: 159.60. Molecular Formula: CuSO4. Color/form: white to pale green pwdr. Strem# 93-2957. http://www.strem.com/catalog/v/93-2957/
Catalog No. 50-901-14906