Learn More
Strem, An Ascensus Company CAS# 7758-99-8. 5g. Copper(II) sulfate pentahydrate (99.999%-Cu) PURATREM. MFCD00149681
SDP
Supplier:  Strem, An Ascensus Company 9329595G
Encompass
CAS# 7758-99-8. 5g. Copper(II) sulfate pentahydrate (99.999%-Cu) PURATREM. MFCD00149681. Molecular Weight: 159.60 (249.68). Molecular Formula: CuSO4∙5H2O. Color/form: blue xtl. Strem# 93-2959. http://www.strem.com/catalog/v/93-2959/
Catalog No. 50-901-14909