Learn More
Hach Company Copper Masking Reagent Powder Pillows, 10 mL, pk/100
Supplier:  Hach Company 2603449
Encompass
For Copper determination by the Porphyrin method. Hach Method 8143 Range: 2 - 210 μg/L Cu. Sample Size: 10 mL Pack of 100 Powder Pillows.
Catalog No. 50-199-6315